siRNA Details
Gene Symbol: ZNF699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198535.2 | NP_940937.1 | 6 | 2080 | ||
GenBank mRNA | AK095463.1 | - | 4 | 1592 | ||
BC109267.1 | - | 5 | 1710 | 40025983 | ||
BC109268.1 | - | 5 | 1710 | 40025986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
