siRNA Details
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 3 | 1456 | ||
XM_005268665.4 | XP_005268722.1 | 3 | 1237 | |||
GenBank mRNA | AY845858.1 | - | 3 | 1317 | ||
BC137413.1 | - | 3 | 1372 | 9021411 | ||
BC137414.1 | - | 3 | 1372 | 9021412 | ||
BC144617.1 | - | 3 | 1346 | |||
DB224774.1 | - | 1 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
