siRNA Details
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 1 | 160 | ||
NM_001308340.1 | NP_001295269.1 | 1 | 160 | |||
GenBank mRNA | AK307955.1 | - | 1 | 115 | ||
AL712776.1 | - | 1 | 153 | |||
AY845858.1 | - | 1 | 21 | |||
BC137413.1 | - | 1 | 76 | 9021411 | ||
BC137414.1 | - | 1 | 76 | 9021412 | ||
BC144617.1 | - | 1 | 76 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
