siRNA Details
Gene Symbol: KIAA1210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020721.1 | NP_065772.1 | 11 | 2216 | ||
XM_006724774.3 | XP_006724837.1 | 9 | 1805 | |||
XM_017029688.1 | XP_016885177.1 | 9 | 1844 | |||
XM_017029689.1 | XP_016885178.1 | 8 | 1646 | |||
GenBank mRNA | AB033036.1 | - | 1 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
