siRNA Details
Gene Symbol: HSPC324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135132.1 | 1 | 31 | |||
GenBank mRNA | AF161442.1 | - | 1 | 32 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
