siRNA Details
Gene Symbol: TEX36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128202.2 | NP_001121674.1 | 3 | 382 | ||
NM_001318133.1 | NP_001305062.1 | 3 | 382 | |||
XM_005269817.4 | XP_005269874.1 | 3 | 380 | |||
GenBank mRNA | AK131010.1 | - | 2 | 55 | ||
BC062717.1 | - | 3 | 385 | 6101709 | ||
BC127766.1 | - | 3 | 263 | 40127344 | ||
BC127767.1 | - | 3 | 267 | 40127345 | ||
BX336391.2 | - | 3 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
