siRNA Details
Gene Symbol: TEX36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128202.2 | NP_001121674.1 | 4 | 800 | ||
GenBank mRNA | AK131010.1 | - | 3 | 473 | ||
BC062717.1 | - | 4 | 803 | 6101709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
