siRNA Details
Gene Symbol: PCNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032223.3 | NP_115599.2 | 6 | 922 | ||
XM_005273987.4 | XP_005274044.1 | 6 | 2806 | |||
XM_005273990.4 | XP_005274047.1 | 6 | 2806 | |||
XM_006718549.3 | XP_006718612.1 | 6 | 2806 | |||
XM_006718551.3 | XP_006718614.1 | 6 | 2806 | |||
XM_011545024.2 | XP_011543326.1 | 7 | 1979 | |||
XM_011545025.2 | XP_011543327.1 | 6 | 2806 | |||
XM_011545028.2 | XP_011543330.1 | 6 | 2806 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
