siRNA Details
Gene Symbol: EHBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099409.1 | NP_001092879.1 | 9 | 1397 | ||
XM_005273867.3 | XP_005273924.1 | 10 | 1436 | |||
XM_005273868.3 | XP_005273925.1 | 10 | 1435 | |||
XM_006718486.3 | XP_006718549.1 | 10 | 1434 | |||
XM_006718487.3 | XP_006718550.1 | 10 | 1434 | |||
XM_017017467.1 | XP_016872956.1 | 10 | 1184 | |||
XM_017017468.1 | XP_016872957.1 | 10 | 1161 | |||
GenBank mRNA | AK024451.1 | - | 5 | 809 | ||
AL713799.1 | - | 1 | 281 | |||
AL834433.1 | - | 9 | 1094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
