siRNA Details
Gene Symbol: SH3RF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099289.2 | NP_001092759.1 | 4 | 1205 | ||
XM_011511109.2 | XP_011509411.1 | 4 | 1307 | |||
XM_011511110.2 | XP_011509412.1 | 2 | 1007 | |||
GenBank mRNA | AK074131.1 | - | 2 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
