siRNA Details
Gene Symbol: NWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144990.1 | NP_001138462.1 | 7 | 3184 | ||
GenBank mRNA | AB033065.2 | - | 1 | 1452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
