siRNA Details
Gene Symbol: THEMIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010923.2 | NP_001010923.1 | 4 | 928 | ||
NM_001164685.1 | NP_001158157.1 | 4 | 928 | |||
NM_001164687.1 | NP_001158159.1 | 4 | 892 | |||
NM_001318531.1 | NP_001305460.1 | 5 | 993 | |||
XM_011535813.2 | XP_011534115.1 | 6 | 1122 | |||
XM_011535814.2 | XP_011534116.1 | 5 | 893 | |||
XM_011535816.1 | XP_011534118.1 | 6 | 1118 | |||
XM_017010848.1 | XP_016866337.1 | 6 | 1122 | |||
XM_017010849.1 | XP_016866338.1 | 4 | 636 | |||
GenBank mRNA | AK124031.1 | - | 4 | 858 | ||
AK128377.1 | - | 5 | 926 | |||
AK292046.1 | - | 4 | 878 | |||
BC130516.1 | - | 4 | 922 | 40146547 | ||
BC144034.1 | - | 4 | 920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
