siRNA Details
Gene Symbol: SUSD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015551.1 | NP_056366.1 | 5 | 2369 | ||
XM_005265034.3 | XP_005265091.1 | 4 | 1917 | |||
XM_017006137.1 | XP_016861626.1 | 5 | 2319 | |||
GenBank mRNA | AB011099.1 | - | 5 | 2369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
