siRNA Details
Gene Symbol: ARL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162491.1 | NP_001155963.1 | 4, 5 | 492 | ||
NR_103708.1 | 4, 5 | 492 | ||||
NR_103709.1 | 4, 5 | 492 | ||||
XM_011530950.2 | XP_011529252.1 | 4, 5 | 1189 | |||
GenBank mRNA | AK301813.1 | - | 4, 5 | 492 | ||
BC140808.1 | - | 4, 5 | 566 | 9021676 | ||
BC144603.1 | - | 4, 5 | 566 | 9053134 | ||
CD300373.1 | - | 1, 2 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
