siRNA Details
Gene Symbol: GRAMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012642.2 | NP_001012660.1 | 12 | 1278 | ||
XM_011521327.2 | XP_011519629.1 | 12 | 1387 | |||
XM_011521328.2 | XP_011519630.1 | 12 | 1738 | |||
XM_011521329.2 | XP_011519631.1 | 12 | 1255 | |||
XM_011521330.2 | XP_011519632.1 | 12 | 1339 | |||
XM_011521331.2 | XP_011519633.1 | 11 | 1221 | |||
XM_011521332.2 | XP_011519634.1 | 13 | 1462 | |||
XM_011521333.2 | XP_011519635.1 | 13 | 1479 | |||
XM_017021997.1 | XP_016877486.1 | 12 | 1422 | |||
XM_017021998.1 | XP_016877487.1 | 12 | 1646 | |||
GenBank mRNA | AK095072.1 | - | 12 | 1268 | ||
AK096501.1 | - | 3 | 797 | |||
AK303724.1 | - | 4 | 686 | |||
BC038451.1 | - | 12 | 1252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
