siRNA Details
Gene Symbol: ZBTB11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024407.1 | 1 | 870 | |||
GenBank mRNA | AK022880.1 | - | 1 | 854 | ||
BC003066.2 | - | 1 | 870 | |||
BC016930.1 | - | 1 | 825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
