siRNA Details
Gene Symbol: LOC105370792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135681.1 | 3 | 565 | |||
GenBank mRNA | AK057625.1 | - | 2 | 2697 | ||
BC022404.1 | - | 1 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
