siRNA Details
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017920.2 | NP_001017920.2 | 4 | 301 | ||
GenBank mRNA | AY324399.1 | - | 4 | 301 | ||
BC127683.1 | - | 4 | 286 | 40127454 | ||
CD675155.1 | - | 4 | 280 | |||
HQ179934.1 | - | 4 | 260 | |||
HQ179935.1 | - | 5 | 379 | |||
HQ179939.1 | - | 3 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
