siRNA Details
Gene Symbol: LRRC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282460.1 | NP_001269389.1 | 3 | 664 | ||
XM_011534989.2 | XP_011533291.1 | 3 | 814 | |||
XM_011534990.2 | XP_011533292.1 | 3 | 814 | |||
XM_011534992.2 | XP_011533294.1 | 3 | 814 | |||
XM_011534993.1 | XP_011533295.1 | 3 | 814 | |||
XM_017020421.1 | XP_016875910.1 | 3 | 814 | |||
XM_017020422.1 | XP_016875911.1 | 2 | 570 | |||
XM_017020423.1 | XP_016875912.1 | 2 | 984 | |||
XM_017020424.1 | XP_016875913.1 | 3 | 814 | |||
XM_017020425.1 | XP_016875914.1 | 3 | 814 | |||
XM_017020426.1 | XP_016875915.1 | 3 | 814 | |||
XM_017020427.1 | XP_016875916.1 | 3 | 814 | |||
XM_017020428.1 | XP_016875917.1 | 3 | 688 | |||
GenBank mRNA | BC030276.1 | - | 3 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
