siRNA Details
Gene Symbol: LRRC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282460.1 | NP_001269389.1 | 9 | 1731 | ||
XM_011534992.2 | XP_011533294.1 | 7 | 1660 | |||
XM_017020421.1 | XP_016875910.1 | 9 | 1881 | |||
XM_017020422.1 | XP_016875911.1 | 8 | 1637 | |||
XM_017020423.1 | XP_016875912.1 | 8 | 2051 | |||
XM_017020428.1 | XP_016875917.1 | 7 | 1534 | |||
XM_017020429.1 | XP_016875918.1 | 8 | 1203 | |||
XM_017020430.1 | XP_016875919.1 | 8 | 1077 | |||
XM_017020431.1 | XP_016875920.1 | 7 | 959 | |||
GenBank mRNA | BC030276.1 | - | 9 | 1726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
