siRNA Details
Gene Symbol: TADA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152293.2 | NP_689506.2 | 2 | 961 | ||
XM_011513595.1 | XP_011511897.1 | 2 | 1641 | |||
GenBank mRNA | AK026299.1 | - | 1 | 129 | ||
AK093974.1 | - | 2 | 834 | |||
AK096655.1 | - | 1 | 1778 | |||
AK126966.1 | - | 2 | 728 | |||
BC047794.1 | - | 2 | 968 | |||
BC101334.1 | - | 3 | 633 | 40025078 | ||
BC101335.1 | - | 3 | 622 | 40025081 | ||
BC101336.1 | - | 3 | 622 | 40025082 | ||
BC101337.2 | - | 3 | 646 | |||
BX641147.1 | - | 1 | 709 | |||
BX648428.1 | - | 1 | 534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
