siRNA Details
Gene Symbol: TADA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152293.2 | NP_689506.2 | 2 | 486 | ||
XM_011513595.1 | XP_011511897.1 | 2 | 1166 | |||
GenBank mRNA | AK093974.1 | - | 2 | 359 | ||
AK096655.1 | - | 1 | 1303 | |||
AK126966.1 | - | 2 | 253 | |||
BC047794.1 | - | 2 | 493 | |||
BC101334.1 | - | 3 | 158 | 40025078 | ||
BC101335.1 | - | 3 | 147 | 40025081 | ||
BC101336.1 | - | 3 | 147 | 40025082 | ||
BC101337.2 | - | 3 | 171 | |||
BX641147.1 | - | 1 | 234 | |||
BX648428.1 | - | 1 | 59 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
