siRNA Details
Gene Symbol: LIPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144869.1 | NP_001138341.1 | 2 | 626 | ||
XM_011545021.2 | XP_011543323.2 | 2 | 926 | |||
GenBank mRNA | CX752266.1 | - | 2 | 608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
