siRNA Details
Gene Symbol: MROH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100878.1 | NP_001094348.1 | 14 | 3057 | ||
XM_011517214.1 | XP_011515516.1 | 14 | 2427 | |||
XM_011517215.1 | XP_011515517.1 | 14 | 2424 | |||
XM_011517216.2 | XP_011515518.1 | 13 | 2359 | |||
XM_011517217.1 | XP_011515519.1 | 12 | 2293 | |||
XM_011517218.1 | XP_011515520.1 | 13 | 2934 | |||
XM_011517221.2 | XP_011515523.1 | 15 | 2192 | |||
XM_011517222.1 | XP_011515524.1 | 14 | 2124 | |||
XM_011517223.1 | XP_011515525.1 | 13 | 2058 | |||
GenBank mRNA | AF086255.1 | - | 1 | 276 | ||
AF289596.1 | - | 3 | 2119 | |||
AK292180.1 | - | 5 | 1705 | |||
BC029031.1 | - | 5 | 1727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
