siRNA Details
Gene Symbol: MROH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100878.1 | NP_001094348.1 | 14 | 2765 | ||
XM_011517218.1 | XP_011515520.1 | 13 | 2642 | |||
GenBank mRNA | AF289596.1 | - | 3 | 1827 | ||
AK292180.1 | - | 5 | 1413 | |||
BC029031.1 | - | 5 | 1435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
