siRNA Details
Gene Symbol: MACROD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080676.5 | NP_542407.2 | 3 | 511 | ||
XM_017027677.1 | XP_016883166.1 | 3 | 535 | |||
GenBank mRNA | AK131348.1 | - | 3 | 642 | ||
DB495787.2 | - | 511 | ||||
DR001087.1 | - | 70 | ||||
EF159161.1 | - | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
