siRNA Details
Gene Symbol: C16orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272051.1 | NP_001258980.1 | 2 | 706 | ||
GenBank mRNA | AB101207.1 | - | 2 | 650 | ||
BC092497.1 | - | 2 | 650 | |||
BQ962326.1 | - | 2 | 707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
