siRNA Details
Gene Symbol: TRAPPC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139444.2 | NP_001132916.1 | 2, 3 | 243 | ||
GenBank mRNA | BX280366.1 | - | 2, 3 | 243 | ||
DY655209.1 | - | 1, 2 | 50 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
