siRNA Details
Gene Symbol: REP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029874.1 | NP_001025045.1 | 1 | 570 | ||
GenBank mRNA | AY662682.1 | - | 1 | 505 | ||
BC140921.1 | - | 1 | 546 | 9056733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
