siRNA Details
Gene Symbol: CBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005189.2 | NP_005180.1 | 5 | 1559 | ||
XM_011525382.1 | XP_011523684.1 | 5 | 1599 | |||
XM_011525383.2 | XP_011523685.1 | 5 | 2561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
