siRNA Details
Gene Symbol: DPY19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015283.1 | NP_056098.1 | 8 | 824 | ||
XM_011515245.2 | XP_011513547.1 | 8 | 973 | |||
XM_011515246.2 | XP_011513548.1 | 8 | 973 | |||
GenBank mRNA | AB020684.1 | - | 5 | 395 | ||
DQ287932.1 | - | 8 | 824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
