siRNA Details
Gene Symbol: ZNF738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027130.1 | 2 | 279 | |||
GenBank mRNA | AK027579.1 | - | 2 | 221 | ||
AK291002.1 | - | 2 | 218 | |||
AK291193.1 | - | 2 | 186 | |||
BC034499.1 | - | 2 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
