siRNA Details
Gene Symbol: HEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020733.1 | NP_065784.1 | 8, 9 | 3258 | ||
XM_005247666.1 | XP_005247723.1 | 9, 10 | 3553 | |||
GenBank mRNA | AB033063.2 | - | 7, 8 | 2853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
