siRNA Details
Gene Symbol: ZNF486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052852.3 | NP_443084.2 | 4 | 639 | ||
GenBank mRNA | AB209550.1 | - | 2 | 342 | ||
BC008936.2 | - | 4 | 623 | |||
BC027608.1 | - | 4 | 644 | |||
BC117268.1 | - | 4 | 579 | 40125819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
