siRNA Details
Gene Symbol: ZNF486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052852.3 | NP_443084.2 | 3 | 333 | ||
GenBank mRNA | AB209550.1 | - | 1 | 36 | ||
BC008936.2 | - | 3 | 317 | |||
BC027608.1 | - | 3 | 338 | |||
BC117268.1 | - | 3 | 273 | 40125819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
