siRNA Details
Gene Symbol: TANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025185.3 | NP_079461.2 | 25 | 6025 | ||
XM_005257203.4 | XP_005257260.1 | 26 | 6252 | |||
XM_006721806.3 | XP_006721869.1 | 28 | 6504 | |||
XM_006721807.3 | XP_006721870.1 | 27 | 6474 | |||
XM_006721811.3 | XP_006721874.1 | 26 | 6373 | |||
XM_011524597.2 | XP_011522899.1 | 28 | 6507 | |||
XM_011524598.2 | XP_011522900.1 | 27 | 6477 | |||
XM_011524599.1 | XP_011522901.1 | 28 | 6333 | |||
XM_011524600.2 | XP_011522902.1 | 27 | 6396 | |||
XM_011524601.2 | XP_011522903.1 | 28 | 6396 | |||
XM_011524602.2 | XP_011522904.1 | 27 | 6285 | |||
XM_017024429.1 | XP_016879918.1 | 27 | 6300 | |||
XM_017024430.1 | XP_016879919.1 | 24 | 9674 | |||
GenBank mRNA | AB032974.2 | - | 1 | 1663 | ||
AB046856.2 | - | 21 | 5626 | |||
AJ278120.1 | - | 12 | 3429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
