siRNA Details
Gene Symbol: TANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025185.3 | NP_079461.2 | 18, 19 | 3274 | ||
XM_005257203.4 | XP_005257260.1 | 19, 20 | 3501 | |||
XM_006721806.3 | XP_006721869.1 | 20, 21 | 3723 | |||
XM_006721807.3 | XP_006721870.1 | 20, 21 | 3723 | |||
XM_006721810.3 | XP_006721873.1 | 20, 21 | 3723 | |||
XM_006721811.3 | XP_006721874.1 | 20, 21 | 3723 | |||
XM_011524597.2 | XP_011522899.1 | 20, 21 | 3723 | |||
XM_011524598.2 | XP_011522900.1 | 20, 21 | 3723 | |||
XM_011524599.1 | XP_011522901.1 | 20, 21 | 3549 | |||
XM_011524600.2 | XP_011522902.1 | 19, 20 | 3612 | |||
XM_011524601.2 | XP_011522903.1 | 20, 21 | 3612 | |||
XM_011524602.2 | XP_011522904.1 | 19, 20 | 3501 | |||
XM_017024429.1 | XP_016879918.1 | 20, 21 | 3549 | |||
XM_017024430.1 | XP_016879919.1 | 16, 17 | 6890 | |||
GenBank mRNA | AB046856.2 | - | 14, 15 | 2875 | ||
AJ278120.1 | - | 4, 5 | 648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
