siRNA Details
Gene Symbol: SLCO4A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024470.1 | 4 | 1129 | |||
GenBank mRNA | BC025345.1 | - | 4 | 1129 | 4862156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
