siRNA Details
Gene Symbol: PIK3R6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010855.3 | NP_001010855.1 | 19 | 2257 | ||
NM_001290211.1 | NP_001277140.1 | 19 | 2625 | |||
NR_110865.1 | 18 | 2352 | ||||
XM_011523677.2 | XP_011521979.1 | 19 | 2151 | |||
XM_011523678.2 | XP_011521980.1 | 19 | 2138 | |||
XM_011523680.2 | XP_011521982.1 | 18 | 2031 | |||
XM_011523681.2 | XP_011521983.1 | 18 | 2402 | |||
XM_011523683.2 | XP_011521985.1 | 16 | 1839 | |||
XM_011523686.2 | XP_011521988.1 | 10 | 1288 | |||
XM_017024228.1 | XP_016879717.1 | 19 | 2153 | |||
XM_017024229.1 | XP_016879718.1 | 20 | 2269 | |||
XM_017024230.1 | XP_016879719.1 | 19 | 2146 | |||
XM_017024231.1 | XP_016879720.1 | 15 | 2330 | |||
GenBank mRNA | AK091819.1 | - | 18 | 2354 | ||
AY753192.1 | - | 17 | 2017 | |||
BC110301.1 | - | 19 | 2608 | |||
BC125169.1 | - | 14 | 1661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
