siRNA Details
Gene Symbol: PIK3R6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010855.3 | NP_001010855.1 | 18 | 2194 | ||
NM_001290211.1 | NP_001277140.1 | 18 | 2562 | |||
NR_110865.1 | 17 | 2289 | ||||
XM_017024229.1 | XP_016879718.1 | 19 | 2206 | |||
XM_017024230.1 | XP_016879719.1 | 18 | 2083 | |||
GenBank mRNA | AK091819.1 | - | 17 | 2291 | ||
AY753192.1 | - | 16 | 1954 | |||
BC110301.1 | - | 18 | 2545 | |||
BC125169.1 | - | 13 | 1598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
