siRNA Details
Gene Symbol: PLAC8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029869.1 | NP_001025040.1 | 1 | 132 | ||
XM_005268381.3 | XP_005268438.1 | 1 | 30 | |||
XM_011537571.2 | XP_011535873.1 | 1 | 30 | |||
GenBank mRNA | BC130586.1 | - | 1 | 118 | 40146617 | |
BC130588.1 | - | 1 | 118 | 40146619 | ||
BC144387.1 | - | 1 | 118 | |||
BC144388.1 | - | 1 | 118 | |||
BC144389.1 | - | 1 | 118 | |||
BC144392.1 | - | 1 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
