siRNA Details
Gene Symbol: PRAMENP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135291.1 | 6 | 2356 | |||
GenBank mRNA | AK131325.1 | - | 6 | 2356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
