siRNA Details
Gene Symbol: FBXO45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105573.1 | NP_001099043.1 | 2 | 678 | ||
GenBank mRNA | AK025697.1 | - | 2 | 406 | ||
BE778263.1 | - | 2 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
