siRNA Details
Gene Symbol: C10orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164375.2 | NP_001157847.1 | 2 | 1527 | ||
NM_001168390.1 | NP_001161862.1 | 2 | 1429 | |||
XM_011539808.2 | XP_011538110.1 | 2 | 2572 | |||
XM_011539809.2 | XP_011538111.1 | 2 | 1395 | |||
GenBank mRNA | AK074172.1 | - | 2 | 1429 | ||
AK123442.1 | - | 2 | 1449 | |||
BC071608.1 | - | 2 | 1527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
