siRNA Details
Gene Symbol: FAM170B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164484.1 | NP_001157956.1 | 2 | 1285 | ||
GenBank mRNA | BC029839.1 | - | 2 | 1273 | ||
BC047597.1 | - | 2 | 1285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
