siRNA Details
Gene Symbol: NIPAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099287.1 | NP_001092757.1 | 6 | 1161 | ||
NM_001172292.1 | NP_001165763.1 | 5 | 1104 | |||
XM_011534552.1 | XP_011532854.1 | 6 | 1073 | |||
GenBank mRNA | AK296972.1 | - | 5 | 1104 | ||
BC105708.1 | - | 5 | 698 | |||
BC105709.1 | - | 5 | 693 | |||
BC105710.1 | - | 5 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
