siRNA Details
Gene Symbol: NIPAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099287.1 | NP_001092757.1 | 6 | 1163 | ||
NM_001172292.1 | NP_001165763.1 | 5 | 1106 | |||
XM_011534552.1 | XP_011532854.1 | 6 | 1075 | |||
GenBank mRNA | AK296972.1 | - | 5 | 1106 | ||
BC105708.1 | - | 5 | 700 | |||
BC105709.1 | - | 5 | 695 | |||
BC105710.1 | - | 5 | 700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
