siRNA Details
Gene Symbol: CTU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012759.2 | NP_001012777.1 | 8 | 884 | ||
NM_001012762.2 | NP_001012780.1 | 8 | 884 | |||
NM_001318507.1 | NP_001305436.1 | 8 | 1097 | |||
NM_001318513.1 | NP_001305442.1 | 7 | 805 | |||
XM_017023210.1 | XP_016878699.1 | 3 | 442 | |||
GenBank mRNA | BC021056.1 | - | 8 | 828 | ||
BC080540.1 | - | 8 | 792 | |||
BC108659.1 | - | 8 | 832 | 6167465 | ||
BC121805.1 | - | 7 | 785 | 40107956 | ||
BC125269.1 | - | 8 | 864 | 40107964 | ||
BC157881.1 | - | 8 | 845 | 8862703 | ||
BM554780.1 | - | 2 | 127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
