siRNA Details
Gene Symbol: CTU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012759.2 | NP_001012777.1 | 5 | 401 | ||
NM_001012762.2 | NP_001012780.1 | 5 | 401 | |||
NM_001318507.1 | NP_001305436.1 | 5 | 401 | |||
NM_001318513.1 | NP_001305442.1 | 4 | 322 | |||
GenBank mRNA | AK299521.1 | - | 361 | |||
BC021056.1 | - | 5 | 345 | |||
BC080540.1 | - | 5 | 309 | |||
BC108659.1 | - | 5 | 349 | 6167465 | ||
BC121805.1 | - | 4 | 302 | 40107956 | ||
BC125269.1 | - | 5 | 381 | 40107964 | ||
BC157881.1 | - | 5 | 362 | 8862703 | ||
BI520274.1 | - | 5 | 401 | |||
BQ430809.1 | - | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
