siRNA Details
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130001.2 | NP_001123473.1 | 4 | 812 | ||
NM_001130002.2 | NP_001123474.1 | 5 | 901 | |||
GenBank mRNA | BC010062.2 | - | 4 | 712 | ||
BC029129.1 | - | 5 | 848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
